Assay Details
| Gene Symbol: LOC101117810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960851.1 | XP_027816652.1 | 13 - 14 | 1404 | 136 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101117810| Gene Symbol: LOC101117810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027960851.1 | XP_027816652.1 | 13 - 14 | 1404 | 136 | |
Set Membership: |
|