Assay Details
| Gene Symbol: TNFSF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012660.4 | XP_004012709.2 | 5 - 6 | 967 | 60 | |
| XM_012185662.3 | XP_012041052.2 | 6 - 7 | 707 | 60 | ||
| XM_027974962.1 | XP_027830763.1 | 5 - 6 | 913 | 60 | ||
| XM_027974964.1 | XP_027830765.1 | 6 - 7 | 636 | 60 | ||
More Information
Set Membership: |
|

TNFSF13
Genomic Map