Assay Details
| Gene Symbol: PLA2G2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012153366.3 | XP_012008756.2 | 2 - 3 | 557 | 92 | |
| XM_027965581.1 | XP_027821382.1 | 2 - 3 | 322 | 92 | ||
| XM_027965582.1 | XP_027821383.1 | 2 - 3 | 1318 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027965581.1, XM_027965582.1 transcripts.
Set Membership: |
|

PLA2G2C
Genomic Map