Assay Details
| Gene Symbol: PLA2G2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012153366.3 | XP_012008756.2 | 3 - 4 | 702 | 59 | |
| XM_027965580.1 | XP_027821381.1 | 2 - 3 | 306 | 59 | ||
| XM_027965581.1 | XP_027821382.1 | 3 - 4 | 467 | 59 | ||
| XM_027965582.1 | XP_027821383.1 | 3 - 4 | 1463 | 59 | ||
More Information
Set Membership: |
|

PLA2G2C
Genomic Map