Assay Details
| Gene Symbol: PPP1R15B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012186934.3 | XP_012042324.3 | 1 - 2 | 2304 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP1R15B| Gene Symbol: PPP1R15B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012186934.3 | XP_012042324.3 | 1 - 2 | 2304 | 113 | |
Set Membership: |
|