Assay Details
| Gene Symbol: NT5C3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098783.2 | XP_014954269.2 | 6 - 7 | 442 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NT5C3B| Gene Symbol: NT5C3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098783.2 | XP_014954269.2 | 6 - 7 | 442 | 68 | |
Set Membership: |
|