Assay Details
Gene Symbol: CFAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957889.1 | XP_027813690.1 | 7 - 8 | 739 | 60 | |
XM_027957890.1 | XP_027813691.1 | 6 - 7 | 609 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
