Assay Details
Gene Symbol: PAK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027971670.1 | XP_027827471.1 | 7 - 8 | 2056 | 60 | |
XM_027971671.1 | XP_027827472.1 | 7 - 8 | 1839 | 60 | ||
XM_027971672.1 | XP_027827473.1 | 6 - 7 | 1780 | 60 | ||
XM_027971673.1 | XP_027827474.1 | 7 - 8 | 2056 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
