Assay Details
| Gene Symbol: ATPSCKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101343.2 | XP_014956829.1 | 2 - 3 | 388 | 67 | |
| XM_027980168.1 | XP_027835969.1 | 2 - 3 | 389 | 67 | ||
| XM_027980169.1 | XP_027835970.1 | 2 - 3 | 392 | 67 | ||
More Information
Set Membership: |
|

ATPSCKMT
Genomic Map