Assay Details
Gene Symbol: RAB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015265.4 | XP_004015314.1 | 6 - 7 | 677 | 60 | |
XM_012190424.3 | XP_012045814.1 | 5 - 6 | 1694 | 60 | ||
XM_012190426.3 | XP_012045816.1 | 5 - 6 | 1416 | 60 | ||
XM_012190427.3 | XP_012045817.1 | 5 - 6 | 1173 | 60 | ||
XM_012190428.3 | XP_012045818.1 | 5 - 6 | 543 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
