Assay Details
Gene Symbol: NFYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012107107.2 | XP_011962497.2 | 7 - 8 | 835 | 52 | |
XM_012107522.2 | XP_011962912.2 | 7 - 8 | 836 | 52 | ||
XM_012107777.2 | XP_011963167.1 | 7 - 8 | 1031 | 52 | ||
XM_012107808.2 | XP_011963198.1 | 8 - 9 | 1092 | 52 | ||
XM_012107839.2 | XP_011963229.1 | 7 - 8 | 844 | 52 | ||
XM_027968399.1 | XP_027824200.1 | 9 - 10 | 1198 | 52 | ||
XM_027968406.1 | XP_027824207.1 | 7 - 8 | 1253 | 52 | ||
XM_027968412.1 | XP_027824213.1 | 7 - 8 | 1253 | 52 | ||
XM_027968414.1 | XP_027824215.1 | 7 - 8 | 1292 | 52 | ||
XM_027968421.1 | XP_027824222.1 | 6 - 7 | 1140 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
