Assay Details
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005886.3 | XP_004005935.3 | 1 - 2 | 152 | 57 | |
XM_004005887.3 | XP_004005936.3 | 1 - 2 | 152 | 57 | ||
XM_012169544.2 | XP_012024934.2 | 1 - 2 | 152 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
