Assay Details
| Gene Symbol: FEZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969116.1 | XP_027824917.1 | 1 - 2 | 796 | 69 | |
| XM_027969117.1 | XP_027824918.1 | 2 - 3 | 646 | 69 | ||
More Information
Set Membership: |
|

FEZF1
Genomic Map