Assay Details
| Gene Symbol: HSD17B8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958662.1 | XP_027814463.1 | 6 - 7 | 674 | 65 | |
| XM_027958663.1 | XP_027814464.1 | 6 - 7 | 659 | 65 | ||
More Information
Set Membership: |
|

HSD17B8
Genomic Map