Assay Details
Gene Symbol: MMRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012179682.3 | XP_012035072.2 | 8 - 9 | 3540 | 69 | |
XM_012179683.3 | XP_012035073.2 | 8 - 9 | 3537 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
