Assay Details
Gene Symbol: CALHM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012182535.3 | XP_012037925.1 | 1 - 2 | 609 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CALHM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012182535.3 | XP_012037925.1 | 1 - 2 | 609 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() |