Assay Details
Gene Symbol: SPART | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012120.4 | XP_004012169.2 | 6 - 7 | 1617 | 64 | |
XM_015098118.2 | XP_014953604.2 | 6 - 7 | 1602 | 64 | ||
XM_027973610.1 | XP_027829411.1 | 6 - 7 | 1549 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
