Assay Details
| Gene Symbol: NMNAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013736.4 | XP_004013785.1 | 3 - 4 | 403 | 70 | |
| XM_015099222.2 | XP_014954708.1 | 3 - 4 | 480 | 70 | ||
| XM_015099223.2 | XP_014954709.1 | 3 - 4 | 321 | 70 | ||
More Information
Set Membership: |
|

NMNAT1
Genomic Map