Assay Details
| Gene Symbol: PDGFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027956338.1 | XP_027812139.1 | 5 - 6 | 1159 | 58 | |
| XM_027956339.1 | XP_027812140.1 | 5 - 6 | 1156 | 58 | ||
More Information
Set Membership: |
|

PDGFC
Genomic Map