Assay Details
Gene Symbol: LOC101110178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027980021.1 | XP_027835822.1 | 2 - 3 | 168 | 108 | |
XM_027980022.1 | XP_027835823.1 | 2 - 3 | 168 | 108 | ||
XM_027980023.1 | XP_027835824.1 | 2 - 3 | 204 | 108 | ||
XM_027980024.1 | XP_027835825.1 | 2 - 3 | 168 | 108 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
