Assay Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958258.1 | XP_027814059.1 | 7 - 8 | 2078 | 73 | |
| XM_027958259.1 | XP_027814060.1 | 7 - 8 | 2092 | 73 | ||
| XM_027958260.1 | XP_027814061.1 | 7 - 8 | 2078 | 73 | ||
More Information
Set Membership: |
|

ASB14
Genomic Map