Assay Details
| Gene Symbol: LOC105616273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967386.1 | XP_027823187.1 | 20 - 21 | 2247 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105616273| Gene Symbol: LOC105616273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027967386.1 | XP_027823187.1 | 20 - 21 | 2247 | 59 | |
Set Membership: |
|