Assay Details
| Gene Symbol: NOX5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012180838.3 | XP_012036228.2 | 14 - 15 | 2166 | 62 | |
| XM_027971525.1 | XP_027827326.1 | 14 - 15 | 2166 | 62 | ||
| XM_027971526.1 | XP_027827327.1 | 14 - 15 | 2163 | 62 | ||
| XM_027971527.1 | XP_027827328.1 | 14 - 15 | 2166 | 62 | ||
More Information
Set Membership: |
|

NOX5
Genomic Map