Assay Details
Gene Symbol: MFHAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015104587.2 | XP_014960073.2 | 1 - 2 | 3062 | 58 | |
XM_027962666.1 | XP_027818467.1 | 1 - 2 | 3062 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
