Assay Details
| Gene Symbol: CEP19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004003012.3 | XP_004003061.1 | 2 - 3 | 348 | 80 | |
| XM_012096467.2 | XP_011951857.1 | 2 - 3 | 345 | 80 | ||
More Information
Set Membership: |
|

CEP19
Genomic Map