Assay Details
| Gene Symbol: OCEL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177718.3 | XP_012033108.2 | 4 - 5 | 1314 | 63 | |
| XM_027969451.1 | XP_027825252.1 | 5 - 6 | 1102 | 63 | ||
More Information
Set Membership: |
|

OCEL1
Genomic Map