Assay Details
| Gene Symbol: CAPSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097064.3 | XP_011952454.1 | 2 - 3 | 275 | 66 | |
| XM_012097065.2 | XP_011952455.1 | 2 - 3 | 273 | 66 | ||
| XM_012097071.3 | XP_011952461.1 | 2 - 3 | 274 | 66 | ||
| XM_027980119.1 | XP_027835920.1 | 2 - 3 | 276 | 66 | ||
| XM_027980120.1 | XP_027835921.1 | 2 - 3 | 277 | 66 | ||
| XM_027980121.1 | XP_027835922.1 | 2 - 3 | 278 | 66 | ||
More Information
Set Membership: |
|

CAPSL
Genomic Map