Assay Details
Gene Symbol: SPSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013743.4 | XP_004013792.1 | 2 - 3 | 893 | 62 | |
XM_012187361.2 | XP_012042751.1 | 2 - 3 | 953 | 62 | ||
XM_012187362.3 | XP_012042752.1 | 2 - 3 | 1625 | 62 | ||
XM_027975666.1 | XP_027831467.1 | 2 - 3 | 1103 | 62 | ||
XM_027975668.1 | XP_027831469.1 | 2 - 3 | 1066 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
