Assay Details
| Gene Symbol: STAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013374.4 | XP_004013423.4 | 3 - 4 | 881 | 55 | |
| XM_027974611.1 | XP_027830412.1 | 3 - 4 | 881 | 55 | ||
More Information
Set Membership: |
|

STAC2
Genomic Map