Assay Details
| Gene Symbol: NFE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006275.4 | XP_004006324.1 | 2 - 3 | 351 | 68 | |
| XM_004006276.4 | XP_004006325.1 | 2 - 3 | 259 | 68 | ||
More Information
Set Membership: |
|

NFE2
Genomic Map