Assay Details
| Gene Symbol: LOC101119820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009624.4 | XP_004009673.2 | 5 - 6 | 557 | 104 | |
| XM_012179515.3 | XP_012034905.2 | 5 - 6 | 530 | 104 | ||
More Information
Set Membership: |
|

LOC101119820
Genomic Map