Assay Details
| Gene Symbol: ERICH6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973915.1 | XP_027829716.1 | 13 - 14 | 1580 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ERICH6B| Gene Symbol: ERICH6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973915.1 | XP_027829716.1 | 13 - 14 | 1580 | 67 | |
Set Membership: |
|