Assay Details
| Gene Symbol: HMBS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016091.4 | XP_004016140.1 | 5 - 6 | 427 | 62 | |
| XM_012095567.3 | XP_011950957.1 | 5 - 6 | 309 | 62 | ||
| XM_012095568.3 | XP_011950958.1 | 5 - 6 | 313 | 62 | ||
| GenBank mRNA | HQ425632.1 | - | 4 - 5 | 237 | 62 | |
More Information
Set Membership: |
|

HMBS
Genomic Map