Assay Details
| Gene Symbol: BIRC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013098.3 | XP_004013147.2 | 1 - 2 | 168 | 95 | |
| XM_027974193.1 | XP_027829994.1 | 1 - 2 | 168 | 95 | ||
More Information
Set Membership: |
|

BIRC5
Genomic Map