Assay Details
| Gene Symbol: C20H6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027959034.1 | XP_027814835.1 | 4 - 5 | 585 | 90 | |
| XM_027959035.1 | XP_027814836.1 | 4 - 5 | 817 | 90 | ||
| XM_027959036.1 | XP_027814837.1 | 3 - 4 | 527 | 90 | ||

C20H6orf89
Genomic Map