Assay Details
| Gene Symbol: CYP24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027976458.1 | XP_027832259.1 | 4 - 5 | 639 | 79 | |
| XM_027976459.1 | XP_027832260.1 | 4 - 5 | 639 | 79 | ||
| GenBank mRNA | EF635857.1 | - | 1 - 2 | 106 | 79 | |
More Information
Set Membership: |
|

CYP24
Genomic Map