Assay Details
| Gene Symbol: ACD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015591.3 | XP_004015640.2 | 5 - 6 | 915 | 72 | |
| XM_012189965.2 | XP_012045355.2 | 5 - 6 | 915 | 72 | ||
| XM_012189966.2 | XP_012045356.2 | 5 - 6 | 894 | 72 | ||
| XM_012189967.2 | XP_012045357.2 | 5 - 6 | 915 | 72 | ||
| XM_012189968.2 | XP_012045358.2 | 5 - 6 | 915 | 72 | ||
| XM_027978094.1 | XP_027833895.1 | 5 - 6 | 915 | 72 | ||
More Information
Set Membership: |
|

ACD
Genomic Map