Assay Details
| Gene Symbol: PRRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107161.3 | XP_011962551.1 | 3 - 4 | 2261 | 61 | |
| XM_027962868.1 | XP_027818669.1 | 3 - 4 | 263 | 61 | ||
| XM_027962869.1 | XP_027818670.1 | 3 - 4 | 247 | 61 | ||
More Information
Set Membership: |
|

PRRG3
Genomic Map