Assay Details
Gene Symbol: KLF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012185.3 | XP_004012234.2 | 3 - 4 | 1205 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
