Assay Details
| Gene Symbol: ADNP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977322.1 | XP_027833123.1 | 4 - 5 | 419 | 67 | |
| XM_027977323.1 | XP_027833124.1 | 5 - 6 | 498 | 67 | ||
| XM_027977324.1 | XP_027833125.1 | 3 - 4 | 223 | 67 | ||
| XM_027977325.1 | XP_027833126.1 | 3 - 4 | 1151 | 67 | ||
| XM_027977326.1 | XP_027833127.1 | 4 - 5 | 1361 | 67 | ||
| XM_027977327.1 | XP_027833128.1 | 3 - 4 | 261 | 67 | ||
| XM_027977328.1 | XP_027833129.1 | 3 - 4 | 429 | 67 | ||
More Information
Set Membership: |
|

ADNP
Genomic Map