Assay Details
| Gene Symbol: CASS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977022.1 | XP_027832823.1 | 4 - 5 | 1461 | 89 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CASS4| Gene Symbol: CASS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977022.1 | XP_027832823.1 | 4 - 5 | 1461 | 89 | |