Assay Details
Gene Symbol: FES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957082.1 | XP_027812883.1 | 17 - 18 | 2346 | 53 | |
XM_027957083.1 | XP_027812884.1 | 17 - 18 | 2328 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() |
