Assay Details
| Gene Symbol: LOC101106044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012162676.3 | XP_012018066.2 | 25 - 26 | 3619 | 61 | |
| XM_027970718.1 | XP_027826519.1 | 25 - 26 | 3619 | 61 | ||
More Information
Set Membership: |
|

LOC101106044
Genomic Map