Assay Details
| Gene Symbol: NDUFB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011650.3 | XP_004011699.2 | 2 - 3 | 349 | 72 | |
| XM_027972913.1 | XP_027828714.1 | 2 - 3 | 226 | 72 | ||
More Information
Set Membership: |
|

NDUFB9
Genomic Map