Assay Details
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027978288.1 | XP_027834089.1 | 2 - 3 | 1130 | 64 | |
XM_027978289.1 | XP_027834090.1 | 1 - 2 | 1463 | 64 | ||
XM_027978290.1 | XP_027834091.1 | 2 - 3 | 1273 | 64 | ||
XM_027978291.1 | XP_027834092.1 | 2 - 3 | 1155 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
