Assay Details
Gene Symbol: LOC101109892 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012186372.3 | XP_012041762.1 | 2 - 3 | 681 | 58 | |
XM_027974315.1 | XP_027830116.1 | 2 - 3 | 229 | 58 | ||
XM_027974316.1 | XP_027830117.1 | 2 - 3 | 220 | 58 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
