Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 10 - 11 | 1519 | 141 | |
XM_012100634.2 | XP_011956024.1 | 6 - 7 | 754 | 141 | ||
XM_027958477.1 | XP_027814278.1 | 9 - 10 | 1491 | 141 | ||
XM_027958478.1 | XP_027814279.1 | 11 - 12 | 1423 | 141 |

More Information
Set Membership: |
![]() ![]() |
