Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 11 - 12 | 1579 | 83 | |
XM_012100634.2 | XP_011956024.1 | 7 - 8 | 814 | 83 | ||
XM_027958477.1 | XP_027814278.1 | 10 - 11 | 1551 | 83 | ||
XM_027958478.1 | XP_027814279.1 | 12 - 13 | 1483 | 83 |
