Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 12 - 13 | 1728 | 69 | |
XM_012100634.2 | XP_011956024.1 | 8 - 9 | 963 | 69 | ||
XM_027958477.1 | XP_027814278.1 | 11 - 12 | 1700 | 69 | ||
XM_027958478.1 | XP_027814279.1 | 13 - 14 | 1632 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
