Assay Details
Gene Symbol: LOC101108001 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100626.3 | XP_011956016.1 | 14 - 15 | 2003 | 84 | |
XM_012100634.2 | XP_011956024.1 | 10 - 11 | 1238 | 84 | ||
XM_027958477.1 | XP_027814278.1 | 13 - 14 | 1975 | 84 | ||
XM_027958478.1 | XP_027814279.1 | 15 - 16 | 1907 | 84 |
